4-Butylbenzoylchlorid ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 4-Butylbenzol-1-carbonylchlorid

A15082.06 A15082.14
A15082.06EA 47.7 EUR
A15082.06 A15082.14
4-Butylbenzoylchlorid ≥98%
4-Butylbenzoylchlorid

Purity: 98% CAS No.: 28788-62-7 Molecular Formula: C11H13ClO Formula Weight: 196.68 Flash Point: 110°(230°F) Refractive Index: 1.5351 Boiling Point: 145-147°/11mm UN No.: 3265 MDL No.: MFCD00000699

Formel: C₁₁H₁₃ClO
Molecular Weight: 196.68 g/mol
Siedepunkt: 145…147 °C (11 mmHg)
Dichte: 1.051
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000699
CAS-Nummer: 28788-62-7
EINECS: 249-224-2
UN: 3265

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR