4-tert-Butylbenzhydrazid

Lieferant: Thermo Scientific

Synonyme: 4-tert-Butylbenzol-1-carbohydrazid, 4-tert-Butylbenzoesäurehydrazid, 4-tert-Butylbenzohydrazid

SPB00257.1 SPB00257.10 SPB00257.50
MAYBSPB00257.1EA 0 EUR
MAYBSPB00257.1 MAYBSPB00257.10 MAYBSPB00257.50
4-tert-Butylbenzhydrazid
4-tert-Butylbenzhydrazid
Formel: C₁₁H₁₆N₂O
Molecular Weight: 192,26 g/mol
MDL: MFCD00014763
CAS-Nummer: 43100-38-5
EINECS: 256-090-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR