2-Nitro-p-toluylsäure

Lieferant: CHEMPUR

BTB05081.1 BTB05081.10
CHMPBTB05081.1EA 0 EUR
CHMPBTB05081.1 CHMPBTB05081.10
2-Nitro-p-toluylsäure
2-Nitro-p-toluylsäure
Formel: C₈H₇NO₄
MDL: MFCD00033899
CAS-Nummer: 27329-27-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR