2-Methoxy-p-toluylsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Methoxy-4-methylbenzoesäure

M-Clarity™ Qualitätsstufe = MQ100

638684-1G 638684-5G
SIAL638684-1GEA 30.5 EUR
SIAL638684-1G SIAL638684-5G
2-Methoxy-p-toluylsäure, Sigma-Aldrich®
2-Methoxy-p-toluylsäure
Formel: C₉H₁₀O₃
Molecular Weight: 166,18 g/mol
MDL: MFCD00020380
CAS-Nummer: 704-45-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR