4-Hydroxy-o-toluylsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Hydroxy-2-methylbenzoic acid, 4-Hydroxy-o-toluic acid

M-Clarity™ Qualitätsstufe = MQ100

653160-10G 653160-1G
SIAL653160-10GEA 258 EUR
SIAL653160-10G SIAL653160-1G
4-Hydroxy-o-toluylsäure, Sigma-Aldrich®
4-Hydroxy-o-toluylsäure
Formel: C₈H₈O₃
Molecular Weight: 152,15 g/mol
MDL: MFCD02182261
CAS-Nummer: 578-39-2
EINECS: 209-422-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR