α,α,α,2,4,6-Hexafluortoluol
Lieferant: CHEMPUR
|
Gefahr
|
Synonyme:
2,4,6-Trifluorobenzotrifluoride, 1,3,5-Trifluoro-2-(trifluoromethyl)benzene
FL5853.5
FL5853.10
CHMPFL5853.5EA
0
EUR
CHMPFL5853.5
CHMPFL5853.10
α,α,α,2,4,6-Hexafluortoluol
α,α,α,2,4,6-Hexafluortoluol
Formel:
C₇H₂F₆ Molecular Weight: 200,08 g/mol Siedepunkt: 105…107 °C (1013 hPa) Dichte: 1,475 g/cm³ (20 °C) |
MDL:
MFCD00190109 CAS-Nummer: 122030-04-0 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...