2-(Trifluormethyl)phenylboronsäure

Lieferant: Thermo Scientific

Synonyme: Dihydroxy(2-(trifluormethyl)phenyl)boran, 2-(Trifluormethyl)benzolboronsäure, (2-(Trifluormethyl)phenyl)metaborsäure

AC35974.250 AC35974.1
MAYBAC35974.250EA 0 EUR
MAYBAC35974.250 MAYBAC35974.1
2-(Trifluormethyl)phenylboronsäure
2-(Trifluormethyl)phenylboronsäure
Formel: C₇H₆BF₃O₂
Molecular Weight: 189,93 g/mol
MDL: MFCD00236059
CAS-Nummer: 1423-27-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR