3-(Trifluormethyl)phenylboronsäure

Lieferant: FLUOROCHEM

011013-5G 011013-25G
FLUO011013-5GEA 45.9 EUR
FLUO011013-5G FLUO011013-25G
3-(Trifluormethyl)phenylboronsäure
3-(Trifluormethyl)phenylboronsäure
Formel: C₇H₆BF₃O₂
MDL: MFCD00151854
CAS-Nummer: 1423-26-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR