(2,3,4,5-Tetrafluorphenyl)acetonitril
Lieferant: FLUOROCHEM
Synonyme:
2,3,4,5-Tetrafluorbenzylcyanid
005847-5G
005847-10G
FLUO005847-5GEA
0
EUR
FLUO005847-5G
FLUO005847-10G
(2,3,4,5-Tetrafluorphenyl)acetonitril
(2,3,4,5-Tetrafluorphenyl)acetonitril
Formel:
C₈H₃F₄N Molecular Weight: 189,11 g/mol Siedepunkt: 123…125 °C (1013 hPa) |
MDL:
MFCD01631454 CAS-Nummer: 53001-74-4 UN: 3439 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...