(2,3,4-Trifluorphenyl)acetonitril

Lieferant: FLUOROCHEM

Synonyme: 2,3,4-Trifluorbenzylcyanid

007696-1G 007696-5G
FLUO007696-1GEA 69.8 EUR
FLUO007696-1G FLUO007696-5G
(2,3,4-Trifluorphenyl)acetonitril
(2,3,4-Trifluorphenyl)acetonitril
Formel: C₈H₄F₃N
Molecular Weight: 171.12 g/mol
MDL: MFCD00061234
CAS-Nummer: 243666-13-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR