(2,4,5-Trifluorphenyl)acetonitril

Lieferant: FLUOROCHEM

007699-1G 007699-5G
FLUO007699-1GEA 34.1 EUR
FLUO007699-1G FLUO007699-5G
(2,4,5-Trifluorphenyl)acetonitril
(2,4,5-Trifluorphenyl)acetonitril
Formel: C₈H₄F₃N
Molecular Weight: 171,12 g/mol
MDL: MFCD00061220
CAS-Nummer: 220141-74-2
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR