(2,4,6-Trifluorphenyl)acetonitril

Lieferant: FLUOROCHEM

Synonyme: 2,4,6-Trifluorbenzylcyanid

007700-1G 007700-5G
FLUO007700-1GEA 72.9 EUR
FLUO007700-1G FLUO007700-5G
(2,4,6-Trifluorphenyl)acetonitril
(2,4,6-Trifluorphenyl)acetonitril
Formel: C₈H₄F₃N
Molecular Weight: 171,12 g/mol
MDL: MFCD00061221
CAS-Nummer: 220227-80-5
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR