(2-Fluorphenyl)acetonitril
Lieferant: FLUOROCHEM
Synonyme:
2-Fluorbenzylcyanid
001187.25G
001187.100G
FLUO001187.25GEA
44.1
EUR
FLUO001187.25G
FLUO001187.100G
(2-Fluorphenyl)acetonitril
(2-Fluorphenyl)acetonitril
Formel:
C₈H₆FN Molecular Weight: 135,14 g/mol Siedepunkt: 114 °C (1013 hPa) Dichte: 1,14045 g/cm³ (15 °C) |
MDL:
MFCD00001897 CAS-Nummer: 326-62-5 EINECS: 206-313-0 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...