(3,5-Difluorphenyl)acetonitril
Lieferant: FLUOROCHEM
Synonyme:
3,5-Difluorbenzylcyanid
004854-5G
004854-25G
FLUO004854-5GEA
34.1
EUR
FLUO004854-5G
FLUO004854-25G
(3,5-Difluorphenyl)acetonitril
(3,5-Difluorphenyl)acetonitril
Formel:
C₈H₅F₂N Molecular Weight: 153.13 g/mol Siedepunkt: 221 Schmelzpunkt: 34…36 °C |
MDL:
MFCD00061278 CAS-Nummer: 122376-76-5 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...