(3-Fluorphenyl)acetonitril
Lieferant: FLUOROCHEM
Synonyme:
3-Fluorbenzylcyanid
001188.5G
001188.25G
001188-100G
FLUO001188.5GEA
0
EUR
FLUO001188.5G
FLUO001188.25G
FLUO001188-100G
(3-Fluorphenyl)acetonitril
(3-Fluorphenyl)acetonitril
Formel:
C₈H₆FN Molecular Weight: 135.14 g/mol Siedepunkt: 113 Dichte: 1.14058 g/cm³ (15 °C) |
MDL:
MFCD00001907 CAS-Nummer: 501-00-8 EINECS: 207-918-2 UN: 3276 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...