2',3'-Difluoracetophenon
Lieferant: FLUOROCHEM
Synonyme:
1-(2,3-Difluorphenyl)ethan-1-on
004775-5G
004775-1G
FLUO004775-5GEA
78.8
EUR
FLUO004775-5G
FLUO004775-1G
2',3'-Difluoracetophenon
2',3'-Difluoracetophenon
Formel:
C₈H₆F₂O |
MDL:
MFCD00061274 CAS-Nummer: 18355-80-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...