3',4'-Difluorpropiophenon

Lieferant: FLUOROCHEM

Synonyme: 1-(3,4-Difluorophenyl)propan-1-one

004874-5G 004874-25G
FLUO004874-5GEA 0 EUR
FLUO004874-5G FLUO004874-25G
3',4'-Difluorpropiophenon
3',4'-Difluorpropiophenon
Formel: C₉H₈F₂O
Molecular Weight: 170.16 g/mol
MDL: MFCD00015505
CAS-Nummer: 23384-72-7
EINECS: 245-627-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR