3',4'-Difluoracetophenon ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-(3,4-Difluorphenyl)ethan-1-on
A15832.06
A15832.14
A15832.22
A15832.06EA
20.8
EUR
A15832.06
A15832.14
A15832.22
3',4'-Difluoracetophenon ≥98%
3',4'-Difluoracetophenon
Purity: 98%
CAS No.: 369-33-5
Molecular Formula: C8H6F2O
Formula Weight: 156.13
Flash Point: 75°(167°F)
Refractive Index: 1.4916
Odour: Characteristic
Melting Point: 19-20°
Boiling Point: 94-95°/13mm
Beilstein Reference: 2206856
MDL No.: MFCD00009891
Formel:
C₈H₆F₂O Molecular Weight: 156,13 g/mol Siedepunkt: 94…95 °C (13 mmHg) Schmelzpunkt: 19…20 °C Dichte: 1,246 Flammpunkt: 75 °C (167 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009891 CAS-Nummer: 369-33-5 EINECS: 206-717-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...