2',6'-Difluoracetophenon ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-(2,6-Difluorphenyl)ethan-1-on

A10220.06 A10220.14
A10220.06EA 90.2 EUR
A10220.06 A10220.14
2',6'-Difluoracetophenon ≥98%
2',6'-Difluoracetophenon

Purity: 98% CAS No.: 13670-99-0 Molecular Formula: C8H6F2O Formula Weight: 156.13 Flash Point: 76°(168°F) Refractive Index: 1.4810 Boiling Point: 76-79°/15mm Beilstein Reference: 2046068 MDL No.: MFCD00000328

Formel: C₈H₆F₂O
Molecular Weight: 156.13 g/mol
Siedepunkt: 76…79 °C (15 mmHg)
Dichte: 1.195
Flammpunkt: 76 °C (168 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000328
CAS-Nummer: 13670-99-0
EINECS: 237-151-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR