2',6'-Difluoracetophenon
Lieferant: FLUOROCHEM
Synonyme:
1-(2,6-Difluorphenyl)ethan-1-on
001954-5G
001954-25G
001954-100G
FLUO001954-5GEA
0
EUR
FLUO001954-5G
FLUO001954-25G
FLUO001954-100G
2',6'-Difluoracetophenon
2',6'-Difluoracetophenon
Formel:
C₈H₆F₂O Molecular Weight: 156,13 g/mol Siedepunkt: 76…79 °C (15 mmHg) Dichte: 1,19 g/cm³ (20 °C) |
MDL:
MFCD00000328 CAS-Nummer: 13670-99-0 EINECS: 237-151-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...