2',5'-Difluoracetophenon
Lieferant: FLUOROCHEM
Synonyme:
1-(2,5-Difluorphenyl)ethan-1-on
001930-10G
001930-25G
001930-5G
FLUO001930-10GEA
0
EUR
FLUO001930-10G
FLUO001930-25G
FLUO001930-5G
2',5'-Difluoracetophenon
2',5'-Difluoracetophenon
Formel:
C₈H₆F₂O Siedepunkt: 70…80 °C (15 mmHg) Dichte: 1,2099 g/cm³ (22 °C) |
MDL:
MFCD00009898 CAS-Nummer: 1979-36-8 EINECS: 217-837-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...