2',3',4',5',6'-Pentafluoracetophenon
Lieferant: FLUOROCHEM
003863-25G
003863-100G
FLUO003863-25GEA
186
EUR
FLUO003863-25G
FLUO003863-100G
2',3',4',5',6'-Pentafluoracetophenon
2',3',4',5',6'-Pentafluoracetophenon
Formel:
C₈H₃F₅O Molecular Weight: 210,1 g/mol Siedepunkt: 130 °C (1013 hPa) Dichte: 1,47 g/cm³ (20 °C) |
MDL:
MFCD00000296 CAS-Nummer: 652-29-9 EINECS: 211-487-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...