2',3',6'-Trifluoracetophenon
Lieferant: FLUOROCHEM
Synonyme:
1-(2,3,6-Trifluorphenyl)ethan-1-on
007470-5G
007470-1G
FLUO007470-5GEA
146
EUR
FLUO007470-5G
FLUO007470-1G
2',3',6'-Trifluoracetophenon
2',3',6'-Trifluoracetophenon
Formel:
C₈H₅F₃O Molecular Weight: 174,12 g/mol |
MDL:
MFCD00070809 CAS-Nummer: 208173-22-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...