2',4',6'-Trifluoracetophenon

Lieferant: FLUOROCHEM

Synonyme: 1-(2,4,6-Trifluorphenyl)ethan-1-on

007471-1G 007471-5G 007471-25G
FLUO007471-1GEA 31.1 EUR
FLUO007471-1G FLUO007471-5G FLUO007471-25G
2',4',6'-Trifluoracetophenon
2',4',6'-Trifluoracetophenon
Formel: C₈H₅F₃O
Molecular Weight: 174,12 g/mol
MDL: MFCD00061194
CAS-Nummer: 51788-77-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR