4'-Fluorpropiophenon ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
p-Fluorpropiophenon
A16219.09
A16219.18
A16219.09EA
27.4
EUR
A16219.09
A16219.18
4'-Fluorpropiophenon ≥97%
4'-Fluorpropiophenon
Purity: 97+%
CAS No.: 456-03-1
Molecular Formula: C9H9FO
Formula Weight: 152.17
Flash Point: 76°(168°F)
Refractive Index: 1.5059
Boiling Point: 100-102°/22mm
Beilstein Reference: 1210310
MDL No.: MFCD00000356
Formel:
C₉H₉FO Molecular Weight: 152,17 g/mol Siedepunkt: 100…102 °C (22 mmHg) Dichte: 1,096 Flammpunkt: 76 °C (168 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000356 CAS-Nummer: 456-03-1 EINECS: 207-255-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...