4'-Fluorpropiophenon
Lieferant: FLUOROCHEM
Synonyme:
p-Fluorpropiophenon
001190.25G
001190.100G
FLUO001190.25GEA
50.1
EUR
FLUO001190.25G
FLUO001190.100G
4'-Fluorpropiophenon
4'-Fluorpropiophenon
Formel:
C₉H₉FO Siedepunkt: 180 °C (1013 hPa) Dichte: 1,09 g/cm³ (20 °C) |
MDL:
MFCD00000356 CAS-Nummer: 456-03-1 EINECS: 207-255-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...