2-Fluor-4-(trifluormethyl)benzoylchlorid

Lieferant: FLUOROCHEM

006403-1G 006403-5G 006403-10G
FLUO006403-1GEA 45.8 EUR
FLUO006403-1G FLUO006403-5G FLUO006403-10G
2-Fluor-4-(trifluormethyl)benzoylchlorid
2-Fluor-4-(trifluormethyl)benzoylchlorid
Formel: C₈H₃ClF₄O
Molecular Weight: 226.56 g/mol
Siedepunkt: 188…189 °C (1013 hPa)
Dichte: 1.5 g/cm³ (20 °C)
MDL: MFCD00061155
CAS-Nummer: 126917-10-0
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR