α,α,α-Trifluor-m-toluoylchlorid

Lieferant: FLUOROCHEM

Synonyme: 3-(Trifluormethyl)benzoesäurechlorid, 3-(Trifluormethyl)benzoylchlorid

007598-25G 007598-100G
FLUO007598-25GEA 39.9 EUR
FLUO007598-25G FLUO007598-100G
α,α,α-Trifluor-m-toluoylchlorid
α,α,α-Trifluor-m-toluoylchlorid
Formel: C₈H₄ClF₃O
Siedepunkt: 184…186 °C (1013 hPa)
Dichte: 1,4078 g/cm³ (20 °C)
MDL: MFCD00000680
CAS-Nummer: 2251-65-2
EINECS: 218-844-5
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR