3-Fluor-4-toluoylchlorid
Lieferant: FLUOROCHEM
Synonyme:
3-Fluor-4-tolylsäurechlorid, 3-Fluor-4-methylbenzoylchlorid
005791-5G
005791-10G
FLUO005791-5GEA
81.8
EUR
FLUO005791-5G
FLUO005791-10G
3-Fluor-4-toluoylchlorid
3-Fluor-4-toluoylchlorid
Formel:
C₈H₆ClFO Molecular Weight: 172,59 g/mol Siedepunkt: 215 °C (1013 hPa) Schmelzpunkt: 38 °C |
MDL:
MFCD00671774 CAS-Nummer: 59189-97-8 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...