3-Fluor-5-(trifluormethyl)benzoylchlorid

Lieferant: FLUOROCHEM

006406-5G 006406-25G
FLUO006406-5GEA 123 EUR
FLUO006406-5G FLUO006406-25G
3-Fluor-5-(trifluormethyl)benzoylchlorid
3-Fluor-5-(trifluormethyl)benzoylchlorid
Formel: C₈H₃ClF₄O
Molecular Weight: 226.56 g/mol
Siedepunkt: 119
Dichte: 1.482 g/cm³ (20 °C)
MDL: MFCD00061157
CAS-Nummer: 171243-30-4
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR