5-Fluor-2-toluoylchlorid

Lieferant: FLUOROCHEM

Synonyme: 5-Fluor-2-tolylsäurechlorid, 5-Fluor-2-methylbenzoylchlorid

006280-5G 006280-1G
FLUO006280-5GEA 171 EUR
FLUO006280-5G FLUO006280-1G
5-Fluor-2-toluoylchlorid
5-Fluor-2-toluoylchlorid
Formel: C₈H₆ClFO
Molecular Weight: 172.59 g/mol
Siedepunkt: 108
Dichte: 1.27 g/cm³ (20 °C)
MDL: MFCD00236681
CAS-Nummer: 21900-39-0
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR