4-Fluor-2-(trifluormethyl)benzoylchlorid
Lieferant: FLUOROCHEM
006407-1G
006407-5G
FLUO006407-1GEA
31.1
EUR
FLUO006407-1G
FLUO006407-5G
4-Fluor-2-(trifluormethyl)benzoylchlorid
4-Fluor-2-(trifluormethyl)benzoylchlorid
Formel:
C₈H₃ClF₄O Molecular Weight: 226.56 g/mol Siedepunkt: 157 Dichte: 1.495 g/cm³ (20 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00061158 CAS-Nummer: 189807-21-4 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...