4-Fluor-3-(trifluormethyl)benzoylchlorid
Lieferant: FLUOROCHEM
006408-1G
006408-5G
FLUO006408-1GEA
37.2
EUR
FLUO006408-1G
FLUO006408-5G
4-Fluor-3-(trifluormethyl)benzoylchlorid
4-Fluor-3-(trifluormethyl)benzoylchlorid
Formel:
C₈H₃ClF₄O Molecular Weight: 226.56 g/mol Siedepunkt: 80…90 °C (1013 hPa) Dichte: 1.493 g/cm³ (20 °C) |
MDL:
MFCD00061159 CAS-Nummer: 67515-56-4 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...