4-(Trifluormethyl)benzamid
Lieferant: CHEMPUR
Synonyme:
4-(Trifluormethyl)benzoesäureamid
SPB01457.250
SPB01457.1
CHMPSPB01457.250EA
0
EUR
CHMPSPB01457.250
MAYBSPB01457.1
4-(Trifluormethyl)benzamid
4-(Trifluormethyl)benzamid
Formel:
C₈H₆F₃NO Molecular Weight: 189,14 g/mol Schmelzpunkt: 250…251 °C |
MDL:
MFCD00007998 CAS-Nummer: 1891-90-3 EINECS: 217-571-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...