4-(Trifluormethyl)benzamid

Lieferant: FLUOROCHEM

Synonyme: 4-(Trifluormethyl)benzoesäureamid

007584-10G 007584-50G
FLUO007584-10GEA 63.9 EUR
FLUO007584-10G FLUO007584-50G
4-(Trifluormethyl)benzamid
4-(Trifluormethyl)benzamid
Formel: C₈H₆F₃NO
Molecular Weight: 189,14 g/mol
Schmelzpunkt: 250…251 °C
MDL: MFCD00007998
CAS-Nummer: 1891-90-3
EINECS: 217-571-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR