2,3-Difluor-p-toluylsäure

Lieferant: FLUOROCHEM

005750-5G 005750-10G 005750-1G
FLUO005750-5GEA 104 EUR
FLUO005750-5G FLUO005750-10G FLUO005750-1G
2,3-Difluor-p-toluylsäure
2,3-Difluor-p-toluylsäure
Formel: C₈H₆F₂O₂
Molecular Weight: 172.13 g/mol
MDL: MFCD01631643
CAS-Nummer: 261763-37-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR