2,6-Difluor-m-toluylsäure

Lieferant: FLUOROCHEM

Synonyme: 2,6-Difluoro-3-methylbenzoic acid

005751-5G 005751-10G 005751-1G
FLUO005751-5GEA 227 EUR
FLUO005751-5G FLUO005751-10G FLUO005751-1G
2,6-Difluor-m-toluylsäure
2,6-Difluor-m-toluylsäure
Formel: C₈H₆F₂O₂
Molecular Weight: 172.13 g/mol
MDL: MFCD01631332
CAS-Nummer: 32890-88-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR