α,α,α,2-Tetrafluor-m-toluylsäure
Lieferant: FLUOROCHEM
CFF631.5
CFF631.10
006394-1G
006394-5G
006394-25G
CHMPCFF631.5EA
268
EUR
CHMPCFF631.5
CHMPCFF631.10
FLUO006394-1G
FLUO006394-5G
FLUO006394-25G
α,α,α,2-Tetrafluor-m-toluylsäure
α,α,α,2-Tetrafluor-m-toluylsäure
Formel:
C₈H₄F₄O₂ Molecular Weight: 208,11 g/mol |
MDL:
MFCD00040980 CAS-Nummer: 115029-22-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...