α,α,α,2-Tetrafluor-p-toluylsäure

Lieferant: FLUOROCHEM

Synonyme: 2-Fluoro-4-(trifluoromethyl)benzoic acid

002014-1G 002014-5G 002014-25G
FLUO002014-1GEA 31.1 EUR
FLUO002014-1G FLUO002014-5G FLUO002014-25G
α,α,α,2-Tetrafluor-p-toluylsäure
α,α,α,2-Tetrafluor-p-toluylsäure
Formel: C₈H₄F₄O₂
Molecular Weight: 208,11 g/mol
Schmelzpunkt: 160 °C
MDL: MFCD00010322
CAS-Nummer: 115029-24-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR