α,α,α,6-Tetrafluor-m-toluylsäure
Lieferant: FLUOROCHEM
Synonyme:
2-Fluoro-5-(trifluoromethyl)benzoic acid
002029-5G
002029-25G
FLUO002029-5GEA
39.9
EUR
FLUO002029-5G
FLUO002029-25G
α,α,α,6-Tetrafluor-m-toluylsäure
α,α,α,6-Tetrafluor-m-toluylsäure
Formel:
C₈H₄F₄O₂ Molecular Weight: 208.11 g/mol |
MDL:
MFCD00061292 CAS-Nummer: 115029-23-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...