α,α,α,6-Tetrafluor-o-toluylsäure

Lieferant: FLUOROCHEM

002046-1G 002046-5G 002046-25G
FLUO002046-1GEA 34.1 EUR
FLUO002046-1G FLUO002046-5G FLUO002046-25G
α,α,α,6-Tetrafluor-o-toluylsäure
α,α,α,6-Tetrafluor-o-toluylsäure
Formel: C₈H₄F₄O₂
Molecular Weight: 208.11 g/mol
MDL: MFCD00040981
CAS-Nummer: 32890-94-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR