α,α,α,3-Tetrafluor-o-toluylsäure

Lieferant: FLUOROCHEM

Synonyme: 3-Fluor-2-(trifluormethyl)benzoesäure

005815-1G 005815-2.5G
FLUO005815-1GEA 130 EUR
FLUO005815-1G FLUO005815-2.5G
α,α,α,3-Tetrafluor-o-toluylsäure
α,α,α,3-Tetrafluor-o-toluylsäure
Formel: C₈H₄F₄O₂
Molecular Weight: 208.11 g/mol
MDL: MFCD01631565
CAS-Nummer: 261951-80-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR