α,α,α,5-Tetrafluor-m-toluylsäure

Lieferant: FLUOROCHEM

Synonyme: 3-Fluor-5-(trifluormethyl)benzoesäure

002159-10G 002159-25G
FLUO002159-10GEA 63.9 EUR
FLUO002159-10G FLUO002159-25G
α,α,α,5-Tetrafluor-m-toluylsäure
α,α,α,5-Tetrafluor-m-toluylsäure
Formel: C₈H₄F₄O₂
Molecular Weight: 208.11 g/mol
MDL: MFCD00061293
CAS-Nummer: 161622-05-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR