α,α,α,5-Tetrafluor-o-toluylsäure

Lieferant: FLUOROCHEM

007912-5G 007912-25G
FLUO007912-5GEA 94.1 EUR
FLUO007912-5G FLUO007912-25G
α,α,α,5-Tetrafluor-o-toluylsäure
α,α,α,5-Tetrafluor-o-toluylsäure
Formel: C₈H₄F₄O₂
Molecular Weight: 208,11 g/mol
MDL: MFCD00083521
CAS-Nummer: 654-99-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR