α,α,α,4-Tetrafluor-o-toluylsäure

Lieferant: FLUOROCHEM

002204-5G 002204-25G
FLUO002204-5GEA 0 EUR
FLUO002204-5G FLUO002204-25G
α,α,α,4-Tetrafluor-o-toluylsäure
α,α,α,4-Tetrafluor-o-toluylsäure
Formel: C₈H₄F₄O₂
Molecular Weight: 208,11 g/mol
Siedepunkt: 251 °C (1013 hPa)
Schmelzpunkt: 121…124 °C
MDL: MFCD00040982
CAS-Nummer: 141179-72-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR