4-(Trifluormethyl)anthranilsäure
Lieferant: FLUOROCHEM
CFF803.
013879-5G
013879-25G
CHMPCFF803.EA
0
EUR
CHMPCFF803.
FLUO013879-5G
FLUO013879-25G
4-(Trifluormethyl)anthranilsäure
4-(Trifluormethyl)anthranilsäure
Formel:
C₈H₆F₃NO₂ Molecular Weight: 205,14 g/mol |
MDL:
MFCD00519334 CAS-Nummer: 402-13-1 EINECS: 206-937-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...