6-(Trifluormethyl)anthranilsäure

Lieferant: FLUOROCHEM

CFF805. 009012-1G 009012-5G
CHMPCFF805.EA 0 EUR
CHMPCFF805. FLUO009012-1G FLUO009012-5G
6-(Trifluormethyl)anthranilsäure
6-(Trifluormethyl)anthranilsäure
Formel: C₈H₆F₃NO₂
Molecular Weight: 205.14 g/mol
MDL: MFCD01569538
CAS-Nummer: 314-46-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR