2-Nitro-α,α,α-trifluor-p-toluylsäure
Lieferant: FLUOROCHEM
Synonyme:
2-Nitro-4-(trifluormethyl)benzoesäure
CFF1250.
005487-25G
005487-100G
CHMPCFF1250.EA
0
EUR
CHMPCFF1250.
FLUO005487-25G
FLUO005487-100G
2-Nitro-α,α,α-trifluor-p-toluylsäure
2-Nitro-α,α,α-trifluor-p-toluylsäure
Formel:
C₈H₄F₃NO₄ Molecular Weight: 235,12 g/mol |
MDL:
MFCD00007142 CAS-Nummer: 320-94-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...