5-Nitro-α,α,α-trifluor-m-toluylsäure ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Nitro-5-(trifluoromethyl)benzoic acid

A18316.06 A18316.14
A18316.06EA 39.3 EUR
A18316.06 A18316.14
5-Nitro-α,α,α-trifluor-m-toluylsäure ≥97%
5-Nitro-α,α,α-trifluor-m-toluylsäure

Purity: 97% CAS No.: 328-80-3 Molecular Formula: C8H4F3NO4 Formula Weight: 235.12 Melting Point: 127-132° Beilstein Reference: 2284651

Formel: C₈H₄F₃NO₄
Molecular Weight: 235,12 g/mol
Schmelzpunkt: 127…132 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00024509
CAS-Nummer: 328-80-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR