5-Nitro-α,α,α-trifluor-m-toluylsäure

Lieferant: FLUOROCHEM

Synonyme: 3-Nitro-5-(trifluoromethyl)benzoic acid

CFF1251.1 006903-5G 006903-25G
CHMPCFF1251.1EA 522 EUR
CHMPCFF1251.1 FLUO006903-5G FLUO006903-25G
5-Nitro-α,α,α-trifluor-m-toluylsäure
5-Nitro-α,α,α-trifluor-m-toluylsäure
Formel: C₈H₄F₃NO₄
Molecular Weight: 235.12 g/mol
MDL: MFCD00024509
CAS-Nummer: 328-80-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR